Safety cleanroom goggles
Both autoclavable and single use goggles have anti-fog systems, optically correct toughened lenses and indirect ventilation systems. They are suitable for those who wear glasses and are 100% latex-free. To ensure the highest quality, every pair of our sterile single use goggles is individually inspected and suitable for medical supplies, personal protection equipment or wherever safety and protective goggles are required.
Product Selection
Safety Goggles
Please choose from a list of products and refine your search by attributes...