Suspended Material
![](/m-ru.vwr.com/ru_RU/images/dp_ohaus_MB23_25.jpg)
VWR offers a variety of products for you to measure water quality from the point of view of suspended materials:
- Rapid method with moisture analysers
- Centrifugation or filtration methods in compliance with European norms
- The method for removing volatile suspended solids from the mainly inorganic substances or fixed solids involves the use of both a furnace and an analytical balance.
Product Selection
![](/m-ru.vwr.com/ru_RU/images/thumb_ohaus_MB23.gif)
Moisture Analysers
Routine moisture analysers...