Cleanroom Stationery
Cleanroom notebooks and cleanroom sticky pads are made from cleanroom paper. The writing on both sides of the sheets possible. We remmonend our durable and resistant clipboards to faciliate the scribble of notes in your cleanrooms and controlled production areas.
Special cleanroom pens with a non-slip grip ease the writing even with cleanroom gloves. The ink (different colors are available) of the cleanroom pens is permanent, also on cold and wet surfaces (e.g. plastic bags, disposable labware) and quick drying. It will not smear but can be scrubbed off.
Paper & Sticky Pads
With minimal particles and ions for cleanroom use, for writing, copying and printing on all common office equipment.
Clipboards
For quick clipping of documents.
Notebooks
Constructed with cleanroom paper for note taking in cleanrooms.
Tapes
Adhesive cleanroom tapes are suitable for color marking, marking of surfaces, equipment, machinery, small parts and they are also suitable for bar code labels.
Pens
Permanent, also on cold and wet surfaces.
Computer accessories
Special immersible and easy to disinfect computer accessories (such as cleanroom keyboards and mouses), which are compatible with all computer systems.