Ultrasonic Baths
Ultrasonic cleaning is the most appropriate process for delicate items.
This cleaning technique uses ultrasound (usually from 15–400 kHz). Efficient cleaning is due to the”Cavitation” phenomenon, which generates in the solution a “cleaning bubble” with very high pressure inside (more than 1000 atm).
Product selection