Everyday use lab consumables
Our VWR Collection range was created for you as an easy choice of essential labware, with a wide selection of quality products that keep your lab running efficiently and effectively. Our consumables including beakers, test tubes, cuvettes, lab notebooks, glassware and more that can be used across a variety of applications. These critical supplies offer quality, value and convenience so you can focus on what is most important – your work.
Product Selection
Bottles
HPDE, PP, PE, glass amber bottles etc. Narrow or wide mouth with tamper-evident seal and screw, snap or dropper caps/closures...
Carboys
Ideal for storage and dispensing of water and other solutions, also media in the lab...
Centrifuge Tubes
PS, PP or PTFE centrifuge tubes, non sterile or sterile, DNase-, RNase-, pyrogen- and TSE-/BSE-free options; available in racks and bags...
Cryogenic Vials
Cryogenic vials for the storage of biological material, human or animal cells at temperatures as low as -196 °C...
Filtration
Filter papers, syringe filters, bottle-top filters etc for chemistry, chromatography and life science filtration...
Microcentrifuge Tubes
Snap-cap and screw cap microcentrifuge tubes from 0,5 to 2,0 ml in a variety of colours...
Microscope Slides and Coverslips
Microscope slides and cover slips/glasses for basic biology teaching to healthcare diagnostics...
Petri Dishes
Gamma irradiated, sterile or asceptic PS dishes in a variety of sizes and formats...
Pipette Tips
Whatever your pipette tip need, VWR have a product that will suit you...
Pipettes, Pasteur/Serological
Plastic or glass transfer/pasteur and serological pipettes in a variety of volumes, formats and packs...
Reservoirs
VWR reagent reservoirs - great for multi-channel work...
Tubes
VWR disposable and reusable test tubes, also tubes for sampling and culture...
Volumetric Flasks
From 5 to 2000 ml with amber options for light-sensitive materials, these VWR volumetric flasks are class A, borosilicate glass with a PP stopper...
Weighing Dishes
Plastic, paper, aluminium or glass, VWR weighing boats, scoops and dishes are available in many volumes down to the micro range, ideal for weighing and moisture determination...