Anti-ITGAL Rat Monoclonal Antibody [clone: 5C28]
Supplier: United States Biological
USBIC2262-03REA 608 EUR
USBIC2262-03R
Anti-ITGAL Rat Monoclonal Antibody [clone: 5C28]
Antibodies
Type: Primary
Antigen: ITGAL (integrin, alpha L (antigen CD11A (p180), lymphocyte function-associated antigen 1; alpha polypeptide))
Clonality: Monoclonal
Clone: 5C28
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse