Anti-FOS Sheep polyclonal antibody unconjugated

Supplier: United States Biological
USBIC0030-05EA 542 EUR
USBIC0030-05
Anti-FOS Sheep polyclonal antibody unconjugated
Antibodies
Type: Primary
Antigen: FOS (FBJ murine osteosarcoma viral oncogene homolog)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Sheep
Isotype:
Reactivity: Hamster, Rabbit, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR