Normal cell line blots, human
Supplier: G-Biosciences
TB53EA 1740 EUR
TB53 TB52 TB51 TB50
Normal cell line blots, human
Electrophoresis Reagents Western Blotting Reagents
Ready to screen, normal tissue human cell line blots from epithelial, endothelial, skin, and muscle tissue
These premade blots are prepared with the appropriate GenLysate™ and 50 μg protein is loaded onto a 4 - 20% denaturing polyacrylamide gel, along with a prestained molecular weight marker. After the proteins are resolved, they are transferred to a PVDF membrane, using western transfer buffer. These blots are ready to be blocked and probed with antibodies of choice.
These premade blots are prepared with the appropriate GenLysate™ and 50 μg protein is loaded onto a 4 to 20% denaturing polyacrylamide gel, along with a prestained molecular weight marker. After the proteins are resolved, they are transferred to a PVDF membrane, using western transfer buffer. These blots are ready to be blocked and probed with antibodies of choice.