Mouse single tissue blot, single species
Supplier: G-Biosciences
TB08EA 624 EUR
TB08 TB17 TB05 TB26 TB14 TB02 TB23 TB11 TB20
Mouse single tissue blot, single species
Electrophoresis Reagents Western Blotting Reagents
This blot contains two protein lanes, a protein marker lane and a lane with one GenLysate™ sample. After the proteins are resolved, they are transferred to a PVDF membrane using Western Transfer Buffer. The blots are ready to be blocked and probed with the antibodies of choice.
  • Ready to screen
  • Single tissue mouse blots