Test strips for Dialysis, DiaQuant®
Supplier: Macherey-Nagel
MANA932004EA 31.1 EUR
MANA932004 MANA932001 MANA932009
Test strips for Dialysis, DiaQuant®
Environmental Test Kits
The DiaQuant® product line is specifically designed for the dialysis environment to test rinse and feed water, as well as sufficient potency levels of disinfectant when storing dialysis equipment. All tests are simple and fast without any mixing or mess. Proven technology makes testing reliable and accurate, thus providing a safe environment for patients.
  • Results within seconds: Just dip, wait and read against multi-stage colour scale
  • Always ready to use: No accessories necessary for evaluation
  • Safe results: Desiccant in the stopper for optimal protection of the strips against humidity
In dialysis, reliable on-site chemical testing is a critical factor for patient safety. Accurate and quick testing is also fundamental to run a dialysis centre effectively and successfully.