Test strips, LubriCheck, QUANTOFIX®
Supplier: MACHEREY-NAGEL
QUANTOFIX®
MANA91336EA 37.5 EUR
MANA91336
Test strips, LubriCheck, QUANTOFIX®
Environmental Test Kits
QUANTOFIX® test strips meet all requirements of a modern rapid test. The colour of the reactive pad changes depending on the concentration of an analyte in the sample. The evaluation is usually carried out visually by comparison of the reaction colour with a multi-stage colour scale. The test strips are intended for single use, maintenance or calibration are not required.
  • Results within seconds: Just dip, wait and read against multi-stage colour scale
  • Always ready to use: No accessories necessary for evaluation
  • Safe results: Desiccant in the stopper for optimal protection of the strips against humidity
These test strips allow the quick and easy determination of the concentration of cooling lubricants. The easy dip-and-read-procedure provides a reliable result within one minute. Cooling lubribants are used when metal parts are being machined (drilling, cutting). Using QUANTOFIX® LubriCheck the concentration of the cooling lubricant can easily be checked on the spot. This ensures optimal cooling and lubrication and at the same time optimal quality of the workpiece.
The measurement with QUANTOFIX® LubriCheck is rapid and reliable and can easily be performed by chemically untrained persons. Other methods used so far either require sensitive instruments (measurement of the refraction index) or have to be performed in the laboratory (determination after digestion). The concentration is detected and given as mmol KOH/L. Using a factor the result can easily be transferred to the concentration of the cooling lubricant.
Gradation: 0 · 15 · 50 · 75 · 130 · 200 mmol/l KOH