Test strips, ammonia
Supplier: Macherey-Nagel
MANA90714EA 22.9 EUR
MANA90714
Test strips, ammonia
Environmental Test Kits
These test strips are for the fast and reliable determination of Ammonium, respectively Ammonia, in solutions. The easy dip-and-read procedure provides safe results within 40 seconds. Ammonium/Ammonia is harmful to fish and other aquatic life in aquariums. This test shows the amount of Ammonium quickly and easily.
  • The aquarium can be kept in good condition and fish stay healthy
  • Results within seconds: Just dip, wait and read against multi-stage colour scale
  • Always ready to use: No accessories necessary for evaluation
  • Safe results: Desiccant in the stopper for optimal protection of the strips against humidity
Always ready to use, no accessories necessary for evaluation. Safe results since the desiccant in the stopper for optimal protection of the strips against humidity.
Colour reaction: Bright yellow to blue