Centrefeed dispenser, WypAll® Reach™
Supplier: Kimberly-Clark
WYPALL®
115-2528EA 50.9 EUR
115-2528 115-2967
Centrefeed dispenser, WypAll® Reach™
Wipe Dispensers
Transparent black, portable, single-sheet wiper dispenser. Ensures rolls remain clean and enclosed to help prevent transmission of food-borne bacteria. Fits service and retail centrefeed rolls (115-2527) and food and hygiene centrefeed rolls (115-2529 and 115-2531).
  • One portable centrefeed white roll or blue roll dispenser
  • Fits one white or blue roll at a time
  • Portable centrefeed roll dispenser designed for use in service, retail, and food preparation environments
  • Design ensures centrefeed rolls remain clean and enclosed to help prevent against transmission of food-borne bacteria
No matter what surfaces they may come into contact with throughout the day, provide employees and visitors greater peace of mind by offering proven cleaning and disinfecting workplace hygiene solutions. WypAll® solutions have been designed to enhance regulatory hygiene practices and compliance, helping to break the chain of germ transmission. WypAll® brand professional wiping paper products are designed for the specific environments where they will be used.
The WypAll® Reach™ centrefeed dispenser is a portable, single-sheet wiper dispenser designed to enable controlled-use of food and hygiene and service and retail centrefeed rolls. This easy to handle, portable and enclosed white roll and blue roll dispenser system is designed to keep the centrefeed roll clean, which helps to prevent against transmission of food-borne bacteria. The single sheet dispensing is designed to minimise contamination whilst encouraging efficient use of the paper wipes. The WypAll® Reach™ dispenser is also available with a bracket which allows housing the dispenser or transforming it into a wall mounted dispenser (115-2528).