VWR®, Nucleic acid loading dyes
Supplier: Avantor
VWR®
E190-5MLEA 49.7 EUR
E190-5ML
VWR®, Nucleic acid loading dyes
Stains Electrophoresis Stains
Designed to optimise loading nucleic acids in agarose gels, the loading dye imparts colour to the sample to facilitate the loading process and increase the density of the sample to ensure efficient sample distribution into each well. The dye migrates independently from the sample, allowing estimation of the migration of nucleic acids.
  • Eliminates exposure to irritating and harmful powdered dyes
  • Free from DNase, RNase and protease
  • Functionally and analytically tested