Sampling siphons
Supplier: Bürkle
BURK5387-0050EA 10.9 EUR
BURK5387-0050 BURK5387-0075 BURK5387-0100
Sampling siphons
Samplers Disposable Samplers
Disposable plunging samplers acting like siphons.
  • Transparent plastic for better visibility
  • PE surface resists adhesion of the material
  • Minimised sample retention on the dip sampler
If strongly adhesive liquids are sampled, it is easier to dispose of the sampler than to carry out a complicated cleaning process.