Burners, Teclu
Supplier: Bochem
BOCH7100EA 138 EUR
BOCH7100 BOCH7110
Burners, Teclu
Heaters Gas Burners
These burners have air regulation and reach a maximum temperature of 1300 °C.
Certifications: DIN 30665