VWR®, GC Rich TEMPase Hot start DNA polymerase and MasterMix
Supplier: Avantor
VWR®
733-2562EA 1940 EUR
733-2562 733-2561 733-2567 733-2565
VWR®, GC Rich TEMPase Hot start DNA polymerase and MasterMix
Nucleic Acid Reagents End-point PCR Enzymes and Kits
VWR® TEMPase Hot Start DNA Polymerases are highly stable polymerases, featuring higher specificity, superior sensitivity and greater yields compared to standard DNA polymerases. These features make them well suited for the detection of low abundance targets. Other uses include screening, amplification of GC-rich sequences, multiplex PCR, direct PCR and qPCR.
The GC-Rich Template kit is specifically designed for difficult GC-rich sequences. Combined with TEMPase, GC buffers I and II promote excellent amplification. The kit is designed for initial testing before using one of the GC-TEMPase 2X Master Mixes.
Ready to use GC TEMPase 2× Master Mixes I and II are designed for amplification of GC-rich sequences.