PerfeCTa® qPCR FastMix™ reaction mixes
Supplier: Quantabio
PerfeCTa® qPCR FastMix™
733-1398EA 1300 EUR
733-1398 733-1395 733-1394 733-1403 733-1402 733-1399
PerfeCTa® qPCR FastMix™ reaction mixes
Nucleic Acid Reagents qPCR/RT-qPCR Enzymes and Kits
FastMixes include buffers and stabilisers optimised for maximum PCR efficiency, reproducibility, and sensitivity with TaqMan® and TaqMan® MGB probe chemistries. A key component of PerfeCTa® qPCR FastMixes is AccuFast™ Taq DNA polymerase that enables instant activation and rapid polymerisation kinetics. Upon heat activation (20 seconds at 95 °C), the antibodies irreversibly denature releasing fully active, unmodified Taq DNA polymerase. This promotes specific and efficient primer extension with the convenience of room temperature reaction assembly. FastMixes deliver maximum PCR efficiency, sensitivity, specificity, and a robust fluorescent signal with either fast or conventional cycling conditions.