VWR®, PCR Plates, 96 Well
Supplier: VWR
VWR®
732-3387EA 180 EUR
732-3387 732-2386 732-3388 732-2387 732-3389 732-2388 732-2389 732-3390 732-2390
VWR®, PCR Plates, 96 Well
Microplates PCR Plates
PP. These PCR plates are compatible with most thermal cyclers.
  • Smooth, thin, uniform well walls ensure accurate thermal transfer
  • Plates are thin and flexible
  • Certified free from DNase, RNase and human genomic DNA
  • Printed alphanumeric labelling and cut corner simplifies plate orientation and sample identification