Educational microscopes for transmitted light observations in routine applications.
- Coated eyepieces, anti-fungus objectives
- Focusing mechanism with limit stop
- Efficient and energy saving LED
- Die-cast frame with high stability and ergonomy
These microscopes have been designed to fulfill all the requirements of an educational laboratory. They give students an effective introduction to the world of microscopy. The quality of the optics and mechanical reliability rank these instruments at the top of their category.
- Achromatic DIN objectives
- Interpupillary adjustment from 48 to 75 mm
- LED illumination with external power supply for low electric consumption
- Focusing mechanism with limit stop to prevent the contact between the objective and specimen
Certifications: Conform EUP 205/32/EC. Achromatic DIN objectives.
Delivery information: Supplied ready to use with eyepieces, objectives, dust cover and manual.