Ideal for laboratory and industrial applications, the durable Scout® STX series features an easy to navigate colour touchscreen display and a slim design which allows stackable storage with optional storage cover.
- Portable if used with 4× AA batteries
- Large, resilient colour touchscreen
- Milligram balances STX123 and STX223 have an ingenious draught shield with an easy to remove top cover piece
- Rapid and efficient navigation with the informative icon driven menu structure
- Fast weighing speed and high resolution deliver repeatable and reliable results
- Overload protection (10× capacity) and transportation lock
- Multiple weighing modes: Weighing, parts counting, % weighing, check weighing, dynamic weighing, totalisation , density determination, display hold and mole weighing
The Scout offers multiple built-in application modes, making even the most complex weighing tasks easy to perform. With nine language options, users can even operate the scale in their own language. A real-time clock function keeps accurate time even during power loss. Additionally, GLP/GMP tracking capabilities assist in the monitoring and reporting of data.
Delivery information: Supplied with AC adaptor, and calibration weights are included with models up to 620 g capacity.