VWR®, Anti-Vibration Weighing Surface
Supplier: VWR
VWR®
611-2536EA 1620 EUR
611-2536 611-5275
VWR®, Anti-Vibration Weighing Surface
Furniture Tables and Workbenches
Polished granite. Four absorbing feet dampen the transmission of extraneous vibrations to this slab making it suitable, not only for high precision weighing, but also microscopy and other similar delicate instrumentation.
Four absorbing feet dampen the transmission of extraneous vibrations to this slab making it suitable, not only for high precision weighing, but also microscopy and other similar delicate instrumentation.