Macrosep® Advance Centrifugal Devices
Supplier: Cytiva (Formerly Pall Lab)
Macrosep®
516-0355EA 323 EUR
516-0355 516-0366 516-0367 516-9919 516-0356 516-0364 516-0365 516-0354 516-0362 516-0483 516-0484 516-0363 516-0360 516-0361 516-0359 516-0357 516-0368 516-0369 516-0358
Macrosep® Advance Centrifugal Devices
Filters Centrifugal Filters
These centrifugal devices can quickly concentrate up to 20 ml of biological sample without valuable sample loss.
  • Provides high recoveries, typically >90%
  • Low protein-binding Omega™ membrane and polypropylene housing minimize losses due to non-specific binding
  • Versatile ultrafiltration Omega™ membrane is available in a variety of MWCOs
  • Built-in deadstop prevents spinning to dryness
  • Color-coded and laser etched for easy identification
Macrosep™ Advance centrifugal devices can be used for a wide range of ultrafiltration applications including the concentration and desalting of proteins, buffer exchange and protein fractionation.
The design of the devices maximizes the effective filtration area allowing for rapid sample processing while maintaining a gentle concentration environment to preserve protein activity and conformation.