Fridge/freezer with spark-free interior for storing explosive and highly flammable substances - for instance in the chemical industry or in special laboratories.
- Comfort electronic controller with digital temperature display enables temperatures to be set accurately
- Integrated data memory
- Dual controllers and dual compressors for optimum functionality
- 1-point calibration
- Access port for temperature sensor (Ø 10 mm)
- Convenient clear drawers (freezer part) with a transparent front providing a clear view of the products stored inside
- The doors are self-closing for convenient use, the door hinge is reversible and the gaskets are replaceable
Tested according to the 94/9 EC (ATEX 95) Directive. Rated as II 3G Ex nA II T6, suitable for storing explosive and highly flammable substances in sealed containers.
Ordering information: Please note that a non spark-free version is also available on request.