Blow lamp, X 2000
Supplier: Camping Gaz
CAMP2000026174EA 69.5 EUR
CAMP2000026174 451-0178
Blow lamp, X 2000
Heaters Gas Burners
Multi-purpose blowtorch for soft and hard soldering and many other applications.
  • Handle made of polypropylene for high impact resistance
  • One-hand operation of the adjustment knob
  • Gas consumption: 140 g/h
  • Flame temperatures up to 1700 °C
Butane cartridge available separately.