Reciprocating shakers, SM 30 B and SM 30 B control, SM 30 C and SM 30 C control
Supplier: Edmund Bühler
444-9322EA 4278 EUR
444-9322 444-9323
Reciprocating shakers, SM 30 B and SM 30 B control, SM 30 C and SM 30 C control
Shakers Reciprocating Shakers
Bench top reciprocating shakers which are suitable for continuous operation and accommodate loads of up to 30 kg. Universal tray accepts max. 11×1000 or 15×500 ml flask clamps; Combifix rack system accommodates 6×1000 or 12×500 ml Erlenmeyer flasks. Variety of different vessels can be accommodated by choosing from the range of optional attachments.
  • SM 30 B has analogue control, SM 30 B control has digital display with a new programmable control and extended functionality
  • Continuous operation or programmable
  • Infinitely variable speed control
  • Can be used at 5 to 50 °C (85% relative humidity), in an incubator or cold room
  • Can be used with illuminated incubator hood TH 30 at ambient +5 to 50 °C with accuracy of ±1% of the set value
Ordering information: Option: Shakers with 46 mm stroke (SM 30 B and SM 30 B control only). Universal tray SM accommodates a maximum 117×10, 64×25, 63×50, 49×100, 23×250, 15×500, 11×1000 or 6×2000 ml flasks or 5 test tube racks. The tray holds 10×100, 6×250, 4×500, 4×1000 or 2×2000 ml separating funnels. Please contact Avantor for details of other accessories that are available.
Delivery information: Supplied with shaking plate and rubber mat, but without universal tray or rack system, accessories must be ordered separately.