VWR®, 4-Place Mini Bead Mill Homogeniser
Supplier: VWR
VWR®
432-0366EA 2650 EUR
432-0366
VWR®, 4-Place Mini Bead Mill Homogeniser
Homogenisers Homogenisers Bench
The VWR® 4-place Mini Bead Mill homogeniser is compact and powerful.
  • Specifically designed for grinding, lysing and the homogenisation of biological samples prior to molecular extraction
  • Ideal solution for releasing DNA, RNA proteins and enzymes from tough samples, while still retaining molecular integrity
  • Offers a broad performance range
Using sample tubes pre-filled with a variety of lysing beads, the mill vigorously and uniformly shakes the tubes providing an efficient, consistent high yield and quality homogenisation in less than 60 seconds. The VWR® 4-place Mini Bead Mill homogeniser supports simultaneous processing of samples in 4×0,5, 4×1,5, 4×2 or 1×7 ml tubes. It does not require a cool down period between runs, allowing non stop use and high throughput. The unit features a convenient front loading design and safety lid closure.
Delivery information: Drive unit is supplied with integrated 0,5, 1,5, and 2 ml tube carriage, interchangeable 1×7 ml tube carriage, user manual and 2 year warranty. VWR® disposable bead tubes must be ordered separately.