Homogeniser, personal, Minilys®
Supplier: Bertin
Minilys®
432-0274EA 2150 EUR
432-0274
Homogeniser, personal, Minilys®
Homogenisers Homogenisers Bench
Especially designed for low throughput laboratory workflows and personal use, Minilys® homogenises 3 samples simultaneously in 0,5/2 ml tubes or one sample in 7 ml tubes using the same 3-D bead beating technology as the Precellys® homogenisers.
  • Small and compact, Minilys® requires minimal bench space
  • Offers optimal mix between efficiency and flexibility
  • Choice of 3 different sizes of Precellys® lysing kits
  • Ready-to-use system with record of last used parameters
Ordering information: For details of the range of available Precellys® lysing and homogenising kits, please contact Avantor.