Sampler, Vampire
Supplier: Bürkle
Vampire
300-1024EA 1068.75 EUR
300-1024
Sampler, Vampire
Samplers Fluid Samplers
The Vampire sampler can be used to take samples from liquids easily, even from deeper depths.
  • Battery-powered hose pump for sampling liquids from depth
  • Easy cleaning and exchangeable hoses prevent impurities
  • Suction depth up to 5 meters
  • Portable sampler is easy to handle and carry
  • Pump attachment is easy to attach to the drive unit
  • Sample bottle is fastened directly onto the sampling instrument by a bottle basket
The portable pump head consists of a mains-independent drive unit, a pump head, and a delivery hose.
The medium being sampled is directed via the closed hose system directly into the sample container. The sample only comes into contact with the hoses and the sample bottle. This excludes the risk of impurities and contamination because the hoses are easy to clean and can also be replaced easily if necessary.
The self-priming peristaltic pump is also suitable for substances that are dirty or contain particles with a maximum viscosity of 500 mPas. The gentle pumping does not change the sample. Thanks to a suction depth of 5 meters, the battery-powered Vampire Sampler easily pumps samples, even from sampling points at lower depths, for example from barrels, tanks, wells, water traps, or wastewater and sewage treatment plants.
The hose pump is perfect for sampling liquid substances in industrial applications such as beverage production or monitoring water quality for environmental protection.
Delivery information: Consisting of a drive unit, pump head, pump hose (30 cm, Marprene), suction hose (5 m, PE), hose connector (PP), sample bottle (250 ml, LDPE), bottle basket, two rechargeable batteries and battery charger.