Peqlab peqGOLD, DNA Ladder Mix
Supplier: VWR
Peqlab peqGOLD
25-2040EA 114 EUR
25-2040
Peqlab peqGOLD, DNA Ladder Mix
Electrophoresis Reagents Molecular Weight Markers
DNA ladder for quantitative and qualitative analysis of dsDNA.
  • Range: 21 fragments, 100 to 10000 bp in size
  • Three highlighted bands for easy orientation
  • Also available as ready to use mix containing Orange G dye instead of bromophenol blue