Laboratory knives
Supplier: RSG Solingen
233-0148EA 168 EUR
233-0148 233-0149 233-0146 233-0147
Laboratory knives
Knives
These ceramic laboratory knives have extremely hard blades which resist chemicals, fractures and corrosion.
  • Anti-magnetic and lightweight
  • Dishwasher-proof
  • Long-life sharpness