VWR®, Carboys, Narrow Neck, Heavy-Duty, Round, with Screw Cap
Supplier: VWR
VWR®
216-1707EA 114 EUR
216-1707 216-1709
VWR®, Carboys, Narrow Neck, Heavy-Duty, Round, with Screw Cap
Bottles Carboys
PP, translucent, with purple screw cap.
  • Excellent chemical resistance
  • Suitable for use from 0 to 135 °C
  • Graduated to identify the volume dispensed
  • Carboys feature an integrated shoulder handle
Use for storing and dispensing solutions and media. Ideal for sterile water storage, polypropylene carboys can be autoclaved before refilling to stop recurring bacterial growth.
Note: Always remove the cap from the carboy before autoclaving.
Certifications: Resin meets US FDA 21 CFR, USP Class VI.