VWR® Critical Swabs
Supplier: VWR
VWR®
149-0339EA 408 EUR
VWRU89031-286 VWRU89031-276 149-0339 VWRU89022-998 VWRU89022-984 149-0333 149-0334 149-0265 149-0343 149-0337 149-0338
VWR® Critical Swabs
Swabs Swabs for Sampling
Swabs feature 100 ppi open or closed cell polyurethane foam, with 100% virgin polypropylene handle. Some have a blue, glass fibre, reinforced polypropylene shaft for extra rigidity, especially for use in critical environments.
  • Non abrasive to protect delicate components
  • Heads are thermally-bonded to the handle without adhesive
  • Withstand most widely used solvents in controlled environments
Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904