Deodorant pads, Odo-Clave®, SP Bel-Art
Supplier: Bel-Art Products, a part of SP
Odo-Clave®
129-4935EA 275 EUR
129-4935 129-4936 129-4937 129-4938
Deodorant pads, Odo-Clave®, SP Bel-Art
Deodorants and Air Fresheners
These pads are impregnated with a deodorant that is released by heat, which doesn't merely mask autoclave odours but actually neutralises them.
  • Protective cover melts in autoclave, allowing deodorant to be heat-activated
  • Easy to handle: simply unwrap and place a pad in each bag prior to autoclaving
  • Available in four pleasant scents
W×L: 32×32 mm
Packaging: Packed 10 pads per bag, 10 bags per box.