Cetrimonium bromide ≥99%, high purity

Supplier: VWR Chemicals
Warning

0833-1KGEA 490 EUR
0833-1KG 0833-500G
Cetrimonium bromide ≥99%, high purity
Cetrimonium bromide

A cationic positively-charged quaternary ammonium compounds. Utilised for applications ranging from solubilising membrane proteins in non-denaturing conditions to creating denaturing conditions during gel electrophoresis.

Formula: CH₃(CH₂)₁₅N(Br)(CH₃)₃
MW: 364,45 g/mol
Melting Pt: 237…243 °C
Flash Pt: 244 °C (closed cup)
MDL Number: MFCD00011772
CAS Number: 57-09-0
UN: 3077
ADR: 9,III
Merck Index: 13,02034

Order Now

Specification Test Results

Purity ≥99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR