Disodium hydrogen phosphate heptahydrate ACS

Supplier: VWR Chemicals

0348-12KGEA 0 EUR
0348-12KG 0348-50KG 0348-1KG 0348-2.5KG 0348-500G
Disodium hydrogen phosphate heptahydrate ACS
Disodium hydrogen phosphate heptahydrate
Formula: HNa₂O₄P·7H₂O
MW: 268,07 g/mol
Melting Pt: 48 °C
Density: 1,7 g/cm³ (20 °C)
MDL Number: MFCD00149180
CAS Number: 7782-85-6

Order Now

Specification Test Results

Chloride <0.001%
Heavy Metals (as Pb) ≤0.001%
Insolubles ≤0.005%
Iron ≤0.001%
pH (5%, Water) at 25 °C 8.7 – 9.3
Purity (as is) 98 – 102%
Sulphate ≤0.005%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR