VWR®, Bovine serum albumin (BSA)

Supplier: VWR
VWR®
0332-100GEA 279 EUR
0332-100G 0332-25G 0332-500G
VWR®, Bovine serum albumin (BSA)
Proteins and Peptides
Heat shock isolated bovine serum albumin (BSA) suitable for use in a wide range of molecular biology applications, including Western blotting.

  • Nuclease-free
  • Protease-free
  • Heavy metals (as Pb) <0,001%
  • Iron <0,0005%
  • Purity >98%
  • Powder form

A standard grade albumin for basic applications, produced by a heat-shock process in caprylic acid.

pH (5%, water) @ 25 °C 6,5 to 7,5.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR