Sodium dodecyl sulphate (SDS) ≥99%, Biotechnology Grade for biotechnology

Supplier: VWR Chemicals

0227-12KGEA 0 EUR
0227-12KG 0227-250G 0227-5KG 0227-1KG 0227-500G 0227-100G
Sodium dodecyl sulphate (SDS) ≥99%, Biotechnology Grade for biotechnology
Sodium dodecyl sulphate (SDS)

For applications ranging from solubilising membrane proteins in non-denaturing conditions to creating denaturing conditions during gel electrophoresis.

Formula: CH₃(CH₂)₁₁OSO₃Na
MW: 288,38 g/mol
Melting Pt: 204…207 °C
Density: 1,1 g/cm³ (20 °C)
Flash Pt: >100 °C
MDL Number: MFCD00036175
CAS Number: 151-21-3
UN: 1325
ADR: 4.1,III
REACH: 01-2119489461-32
Merck Index: 13,08710
https://export.vwr.com/stibo/web/std.lang.all/54/91/735491.jpg

Order Now

Specification Test Results

Abs.@260nm (3%, Water) <0.1
Abs.@280nm (3%, Water) ≤0.1
Chloride ≤0.1%
Copper ≤0.0005%
DNase none detected
Heavy Metals (as Pb) ≤0.0005%
Iron ≤0.0001%
Phosphate ≤0.0005%
Protease none detected
Purity ≥99%
RNase none detected
Solubility (10%, Water) Pass

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR