Glycine ≥99% for biotechnology

Supplier: VWR Chemicals

0167-12KGEA 637 EUR
0167-12KG 0167-1KG 0167-10KG 0167-5KG 0167-50KG
Glycine ≥99% for biotechnology
Glycine
Formula: NH₂CH₂COOH
MW: 75,07 g/mol
Boiling Pt: 181 °C (1013 hPa)
Melting Pt: 83 °C
Density: 1,64 g/cm³ (20 °C)
MDL Number: MFCD00008131
CAS Number: 56-40-6
Merck Index: 13,04504
https://export.vwr.com/stibo/web/std.lang.all/19/10/2301910.jpg

Order Now

Specification Test Results

Abs.@260 nm (1M solution) ≤0.1
Abs.@280 nm (1M solution) ≤0.1
Chloride ≤0.01%
DNase none detected
Heavy Metals (as Pb) ≤0.002%
Loss on Drying ≤0.2%
pH (5%, Water) @25 °C 5.9 - 6.4
Protease none detected
Purity ≥99%
RNase none detected
Solubility (10%, Water) Pass
Sulphate ≤0.05%
Thin Layer Chromatography (TLC) One Spot

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR