Magnesium acetate tetrahydrate
Supplier: VWR Chemicals
0131-500GEA 81.6 EUR
0131-500G
Magnesium acetate tetrahydrate
Magnesium acetate tetrahydrate
Formula: (H₃CCOO)₂Mg·4H₂O
MW: 214,46 g/mol
Melting Pt: 80 °C
Density: 1,454 g/cm³ (20 °C)
MDL Number: MFCD00149214
CAS Number: 16674-78-5
Order Now
Specification Test Results
Chloride ≤0.01%
Insolubles ≤0.005%
Iron ≤0.01%
Purity
Sulphate ≤0.05%