Compact temperature and humidity logger
Supplier: Rotronic
620-2364EA 137 EUR
620-2364
Compact temperature and humidity logger
Data Loggers
This compact logger with high level of IP protection provides a large storage capacity and maximum/minimum/average statistical function. Internal humidity sensor and NTC temperature sensor.
  • High accuracy
  • Clear LCD display with visual alerts
  • HW4-lite PC software for programming and analysing
  • High measurement accuracy of ±3,0% RH and ±0,3 °C
  • Compact design with a high IP protection
  • MIN/MAX/AVG statistical function
Large storage capacity: 32000 lines, programmable start and stop time with the HW4 software.
Certifications: IP 67 rated enclosure. Conformity to FDA CFR 21 Part 11 /GAMP 5.
Delivery information: Set includes CR2 long life battery, function and calibration certificate and HW4-lite software key code.