VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: Avantor
VWR® AVS TITRINORM®
32033.374EA 174 EUR
32033.374 32095.184 85680.295 32044.268 32095.264 32033.294 32041.292 932045.262 32096.267 32032.291 32096.187 32042.295 85680.290 83601.290 98192.260 98213.260 32097.261 32031.297 32044.372 32044.418 32095.366BOM 32039.374 32044.292 32044.290 32039.294 32097.290 32046.298 32045.290 32038.291 32046.378 98192.290 32096.400 32044.292BOM1 83601.260 32045.375 32044.372BOM 32040.185 32040.260 32045.295 32096.360 83610.610 32045.412 32039.261 32095.366 32035.291 932045.375 32095.297 32046.265 932045.295 32032.260 32040.378 32040.298 32036.294 32096.291 32044.292BOM2 32045.262 32096.371 83610.600 32095.377 85680.260
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Buffers pH Calibration Buffers
Solutions prepared from AnalaR® NORMAPUR® grade analytical reagents.
  • Reference temperature 20 °C
  • True analytical value on the label to an accuracy of ±0,02 pH units (pH 1,00 to 11,00); ±0,05 for pH 12,00 buffer
  • Traceable to SRM from NIST
  • Batch number and packing date for traceability
  • Certificate of Analysis available at website
Manufactured from analytical grade products presenting high accuracy: 0,02 pH unit from pH 1,00 to 11,00 and 0,05 pH unit for pH 12,0.
Certifications: Certifications: Products with accreditation to ISO 17034 in the table below are produced and certified in accordance with ISO 17034 accredited by BELAC under the certification number 542-RM. All information regarding the use of these CRMs can be found on the Certificate of Analysis.
Ordering information: If significantly large volumes are required please contact your local Avantor sales office for a customised offer.
Packaging: Available in different pack sizes: 100 ml, 500 ml, 1 L, 5 L plastic bottles and bag-in-box (5 and 10 L).