VWR® Lens Cleaning Tissues, Grade 541
Supplier: VWR
VWR®
111-5004EA 130 EUR
111-5004 111-5003
VWR® Lens Cleaning Tissues, Grade 541
Wipes Disposable Wipes and Towels Lens Papers
Suitable for cleaning various optical lenses such as objectives, binoculars, magnifying glasses, spectacles, as well as optical fibre filaments.