VWR Funnel family
NEW!
VALID UNTIL: 30/11/2012
  • PP Analytical funnels
    These funnels are designed for use with standard filter papers. The funnels have a 60° body angle and a 58° internal rib angle for rapid filtration.
        - External ribbing prevents airlock
        - Funnels resist most chemicals except strong oxidising agents
        - Can be used from 0 to 135 °C (32 to 275 °F)
        - Autoclavable, PP natural colour
  • PP Powder funnels
    These funnels have a 60° body angle and a 58° internal rib angle for rapid filtration.
        - External ribbing prevents airlock
        - Funnels resist most chemicals except strong oxidising agents
        - May be used from 0 to 135 °C (32 to 275 °F)
        - Autoclavable, natural polypropylene
  • Single use funnels 
        - Polypropylene or polystyrene disposable funnel
        - For use with filter paper or on its own
        - Tapered at a 60° angle with inside fluting
        - Stem diameter top/bottom - 7.4/4.3 mm
Please wait while loading...
Size (mm) Funnel outer ø (mm) Stem ø (mm) Stem height (mm) Total height (mm) Pk Cat.No. Promo Price Qty.
PP analytical funnels
35 41,1 6,3 49,3 75,8 12 221-0243P
45 50,3 7,1 52 84,6 12 221-0244P
55 61,5 7,1 59,5 103,4 12 221-0245P
65 71,1 6,4 61,4 111,7 12 221-0246P
75 83,2 6,8 76,3 139,4 6 221-0247P
90 98,5 8,2 89 164 6 221-0248P
100 108,7 9,1 95,4 180 4 221-0249P
160 168 14 154 280 2 221-0250P
PP powder funnels
65 70 15,6 20,7 66,7 12 221-0239P
80 84,8 15,8 30 85,2 12 221-0240P
100 106,5 20,5 33 106,2 6 221-0241P
150 149,4 26,7 33 136,5 4 221-0242P
PP Single use funnels
65 68 7,4 22,7 74,4 100 221-0251P
55 60 7,4 27,1 71,3 100 221-0252P
PS Single use funnels
55 60 7,4 27,1 71,3 100 221-0253P